accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClOHONON

CAS RN 145645-62-1

IUPAC Name: hydrogen 1-(2-{[(diphenylmethylidene)amino]oxy}ethyl)-1,2,5,6-tetrahydropyridine-3-carboxylic acid chloride
Synonyms: hydrogen 1-(2-{[(diphenylmethylidene)amino]oxy}ethyl)-5,6-dihydro-2H-pyridine-3-carboxylic acid chloride
Molecular Weight (g/mol): 386.88
Molecular Formula: C21H23ClN2O3
InChi Key: YZYRTEYMUTWJPL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)C1=CCCN(CCON=C(C2=CC=CC=C2)C2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results