CAS RN 145821-59-6
IUPAC Name:
hydrogen (3R)-1-[4,4-bis(3-methylthiophen-2-yl)but-3-en-1-yl]piperidine-3-carboxylic acid chloride
Synonyms:
hydrogen tiagabine chloride
Molecular Weight (g/mol):
412
Molecular Formula:
C20H26ClNO2S2
InChi Key:
YUKARLAABCGMCN-PKLMIRHRSA-N
SMILES:
[H+].[Cl-].CC1=C(SC=C1)C(=CCCN1CCC[C@H](C1)C(O)=O)C1=C(C)C=CS1
1
–
1
of
1
results
1
R&D Systems™ Tiagabine hydrochloride
CAS: 145821-59-6 Molecular Formula: C20H26ClNO2S2