CAS RN 145858-50-0
IUPAC Name:
hydrogen 6-[(3-chlorophenyl)(1H-imidazol-1-yl)methyl]-1H-1,3-benzodiazole chloride
Synonyms:
liarozole
Molecular Weight (g/mol):
345.23
Molecular Formula:
C17H14Cl2N4
InChi Key:
OTSZCHORPMQCBZ-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].ClC1=CC=CC(=C1)C(N1C=CN=C1)C1=CC=C2N=CNC2=C1
1
–
1
of
1
results
1
R&D Systems™ Liarozole hydrochloride
CAS: 145858-50-0 Molecular Formula: C17H14Cl2N4