CAS RN 14592-89-3
IUPAC Name:
2,8,15-trimethyl-4,10,13-tris(trifluoromethyl)-1λ³,5,7λ³,11,12λ³,16-hexaoxa-6-chromaspiro[5.5⁶.5⁶]hexadeca-1,3,7,9,12,14-hexaene
Synonyms:
2,8,15-trimethyl-4,10,13-tris(trifluoromethyl)-1λ³,5,7λ³,11,12λ³,16-hexaoxa-6-chromaspiro[5.5⁶.5⁶]hexadeca-1,3,7,9,12,14-hexaene
Molecular Weight (g/mol):
514.26
Molecular Formula:
C15H15CrF9O6
InChi Key:
YPJYEKCGHXZCQF-RQQUGGEBSA-N
SMILES:
[H][O]1=C(C)C=C(O[Cr]1123OC(C)=CC(=[O]1[H])C(F)(F)F)C(F)(F)F.[H][O]2=C(C)C=C(O3)C(F)(F)F
1
–
1
of
1
results
1
Tris(trifluoro-2,4-pentanedionato)chromium(III) 98.0+%, TCI America™
CAS: 14592-89-3 Molecular Formula: C15H15CrF9O6 Molecular Weight (g/mol): 514.26 MDL Number: MFCD00059456 InChI Key: YPJYEKCGHXZCQF-RQQUGGEBSA-N Synonym: Chromium(III) Trifluoroacetylacetonate, Trifluoroacetylacetono Chromium(III) Salt