accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 145965-14-6

IUPAC Name: (4-bromonaphthalen-1-yl)boronic acid
Synonyms: 4-bromonaphthalen-1-ylboronic acid
Molecular Weight (g/mol): 250.89
Molecular Formula: C10H8BBrO2
InChi Key: DKKOMUVRCPKTFX-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(Br)C=C1
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)