accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHOHNNONHONH

CAS RN 146-80-5

IUPAC Name: 9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,3,6,9-tetrahydro-1H-purine-2,6-dione
Synonyms: xanthosine
Molecular Weight (g/mol): 284.23
Molecular Formula: C10H12N4O6
InChi Key: UBORTCNDUKBEOP-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=NC2=C1NC(=O)NC2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results