accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 1460-18-0

IUPAC Name: pentadecanedioic acid
Synonyms: pentadecanedioic acid
Molecular Weight (g/mol): 272.39
Molecular Formula: C15H28O4
InChi Key: BTZVDPWKGXMQFW-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCCC(O)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)