Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecanedioic Acid 97.0+%, TCI America™
SDP

Catalog No. P09091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09091G 1 g
P090910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09091G Supplier TCI America Supplier No. P09091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1460-18-0
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.39
MDL Number MFCD00039534
InChI Key BTZVDPWKGXMQFW-UHFFFAOYSA-N
Synonym 1,15-pentadecanedioic acid, pubchem18101, 1,15-pentadecanoic acid, 1,15-pentadecanedioicacid, acmc-1c0t0, ksc180a4d
PubChem CID 160576
IUPAC Name pentadecanedioic acid
SMILES OC(=O)CCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 272.39
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentadecanedioic Acid
EINECSNumber (2)-3488
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.