Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecanedioic acid, 97%, Thermo Scientific™

Catalog No. AAL0989103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0989103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0989103 Supplier Thermo Scientific Chemicals Supplier No. L0989103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store in cool dry condition. Incompatib
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-18-0
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.39
MDL Number MFCD00039534
InChI Key BTZVDPWKGXMQFW-UHFFFAOYSA-N
Synonym 1,15-pentadecanedioic acid, pubchem18101, 1,15-pentadecanoic acid, 1,15-pentadecanedioicacid, acmc-1c0t0, ksc180a4d
PubChem CID 160576
IUPAC Name pentadecanedioic acid
SMILES OC(=O)CCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 113°C to 114°C
Boiling Point 212°C (16 mmHg)
Quantity 1 g
Beilstein 1711722
Solubility Information Insoluble in water.
Formula Weight 272.39
Percent Purity 97%
Chemical Name or Material Pentadecanedioic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 679-810-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.