accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)NH3OOOO

CAS RN 14603-76-0

IUPAC Name: (2R)-2-azaniumyl-2-methylbutanedioate
Synonyms: (2R)-2-ammonio-2-methylbutanedioate
Molecular Weight (g/mol): 146.12
Molecular Formula: C5H8NO4
InChi Key: CWAYDJFPMMUKOI-RXMQYKEDSA-M
SMILES: C[C@@]([NH3+])(CC([O-])=O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)