Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-alpha-Methylaspartic acid, 98%, 98% ee, Thermo Scientific™

Catalog No. AC290142500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290142500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290142500 Supplier Thermo Scientific Chemicals Supplier No. 290142500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14603-76-0
Molecular Weight (g/mol) 147.13
InChI Key CWAYDJFPMMUKOI-RXMQYKEDSA-N
Synonym 2-methyl-d-aspartic acid, alpha-methyl-d-asp, r---2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl, 2r-2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl-, hydrochloride 1:1, ambotzhaa7540, a-methyl-d-asp, h-a-me-d-asp-oh, r-2-methylaspartic acid
PubChem CID 6993662
IUPAC Name (2R)-2-amino-2-methylbutanedioic acid
SMILES CC(CC(=O)O)(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Enantiomeric Excess > 75%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H9NO4
Linear Formula HOOCC(CH3)nH2CH2COOH
Quantity 250 mg
Specific Rotation -46.8° to -54.8° (25°C, 589nm) (c=0.14 in water)
Formula Weight 147.13
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material (R)-α-Methylaspartic acid, 99+%, (80% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.