accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OCH3

CAS RN 14620-52-1

IUPAC Name: 1,1-dimethoxydodecane
Synonyms: lauraldehyde dimethyl acetal
Molecular Weight (g/mol): 230.39
Molecular Formula: C14H30O2
InChi Key: AJUWUYJULVYGRA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results