Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecanal Dimethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. D193725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D193725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D193725ML Supplier TCI America Supplier No. D193725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14620-52-1
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00036645
InChI Key AJUWUYJULVYGRA-UHFFFAOYSA-N
Synonym Lauryl Aldehyde Dimethyl Acetal, 1,1-Dimethoxydodecane
PubChem CID 84559
IUPAC Name 1,1-dimethoxydodecane
SMILES CCCCCCCCCCCC(OC)OC

Specifications

Color Colorless
Boiling Point 133°C
Quantity 25 mL
Formula Weight 230.39
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecanal Dimethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.