accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNN

CAS RN 146406-75-9

IUPAC Name: 6-(pyridin-2-yl)-4-{4-[6-(pyridin-2-yl)-[2,2'-bipyridin]-4-yl]phenyl}-2,2'-bipyridine
Synonyms: 6-(pyridin-2-yl)-4-{4-[6-(pyridin-2-yl)-[2,2'-bipyridin]-4-yl]phenyl}-2,2'-bipyridine
Molecular Weight (g/mol): 540.63
Molecular Formula: C36H24N6
InChi Key: NXMHYAQTBVTLRK-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=CC(=CC(=N1)C1=CC=CC=N1)C1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results