accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONNO

CAS RN 14663-53-7

IUPAC Name: 2-(4-oxo-3,4-dihydroquinazolin-3-yl)acetate
Synonyms: 2-(4-oxoquinazolin-3-yl)acetate
Molecular Weight (g/mol): 203.18
Molecular Formula: C10H7N2O3
InChi Key: BBBICQPTZBHLBM-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1C=NC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results