CAS RN 1467057-23-3
CAS RN 1467057-23-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ PF 06409577

Potent and selective allosteric activator of AMPK