CAS RN 147-85-3
L-Proline, 99%
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: (2S)-pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
L-Proline, 99%, MP Biomedicals, MP Biomedicals
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
L-Proline, >99%, MP Biomedicals™
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
L-Proline, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
L-Proline, FCC, 98.5-101.5%, Spectrum™ Chemical
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
phyproof™ L-Proline, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Proline, USP, 98.5-101.5%, Spectrum™ Chemical
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
L-Proline 99.0+%, TCI America™
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
Fisher Scientific L-Proline (White Crystals or Crystalline Powder), Fisher BioReagents
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1
L-Proline, Multi-Compendial, U.S.P., J.T. Baker™
CAS: 147-85-3 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 MDL Number: MFCD00064318 InChI Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N Synonym: l-proline,proline,l---proline,s-pyrrolidine-2-carboxylic acid,2-pyrrolidinecarboxylic acid,--proline,--s-proline,2s-pyrrolidine-2-carboxylic acid,prolinum,h-pro-oh PubChem CID: 145742 ChEBI: CHEBI:17203 IUPAC Name: (2S)-pyrrolidine-2-carboxylic acid SMILES: OC(=O)C1CCCN1