Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Proline (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP392100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP392-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP392-100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

Melting Point 228°C
Ammonium 0.02% max.
pH 5.5 to 7.0
Color White
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.3% max.
Packaging Glass Bottle
Residue after Ignition 0.1% max.
Specific Rotation -84.5 to -86.0° (+ or -)
Merck Index 15, 7895
State of Solution (%T) 98.0% min.
Alpha Vector Proline
Density 1.350g/cm³
Quantity 100 g
Formula Weight 115.13
Percent Purity 98.5 to 101.0%
Physical Form Powder
Chemical Name or Material L-Proline
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May be harmful by inhalation, in contact with skin and if swallowed. Avoid contact with skin and eyes. Avoid dust formation in confined areas. Do not breathe dust. Do not breathe vapors or spray mist. Do not ingest. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Remove from exposure, lie down. Move to fresh air. Clean mouth with water. Get medical attention. Get medical attention. .

NFPA

Health:1
Flammability:1
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.