CAS RN 147416-96-4
IUPAC Name:
dihydrogen 4-methyl-2-[2-(4-methylpiperazin-1-yl)acetyl]-5-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3,6,10,12-pentaen-8-one dichloride
Synonyms:
dihydrogen telenzepine dichloride
Molecular Weight (g/mol):
443.39
Molecular Formula:
C19H24Cl2N4O2S
InChi Key:
JTQJFSQQHGPLOX-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CN1CCN(CC(=O)N2C3=C(C)SC=C3C(=O)NC3=CC=CC=C23)CC1
1
–
1
of
1
results
1
Tocris Bioscience™ Telenzepine dihydrochloride
CAS: 147416-96-4 Molecular Formula: C19H24Cl2N4O2S