accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFOHOOH

CAS RN 14742-36-0

IUPAC Name: 2,3,4,5-tetrafluoro-6-hydroxybenzoic acid
Synonyms: 2,3,4,5-tetrafluoro-6-hydroxybenzoic acid
Molecular Weight (g/mol): 210.08
Molecular Formula: C7H2F4O3
InChi Key: WKYGDMSOKGXAAY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results