accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOONHBr

CAS RN 147523-65-7

IUPAC Name: N-(4-bromophenyl)-3-oxo-4,5-diphenylpyrazolidine-1-carboxamide
Synonyms: N-(4-bromophenyl)-3-oxo-4,5-diphenylpyrazolidine-1-carboxamide
Molecular Weight (g/mol): 436.31
Molecular Formula: C22H18BrN3O2
InChi Key: LMUQHXHWJWQXSD-UHFFFAOYNA-N
SMILES: BrC1=CC=C(NC(=O)N2NC(=O)C(C2C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)