CAS RN 147526-32-7
IUPAC Name:
calcium bis((3R,5S,6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate)
Synonyms:
calcium bis((3R,5S,6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate)
Molecular Weight (g/mol):
881
Molecular Formula:
C50H46CaF2N2O8
InChi Key:
RHGYHLPFVJEAOC-FFNUKLMVSA-L
SMILES:
[Ca++].O[C@H](C[C@H](O)\C=C\C1=C(N=C2C=CC=CC2=C1C1=CC=C(F)C=C1)C1CC1)CC([O-])=O.O[C@H](C[C@H](O)\C=C\C1=C(N=C2C=CC=CC2=C1C1=CC=C(F)C=C1)C1CC1)CC([O-])=O
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Pitavastatin calcium
Thermo Scientific Chemicals Pitavastatin calcium
CAS: 147526-32-7 Molecular Formula: C50H46CaF2N2O8
2
R&D Systems™ Pitavastatin calcium
CAS: 147526-32-7 Molecular Formula: C50H46CaF2N2O8