accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NOHNOCH3

CAS RN 1476-98-8

IUPAC Name: hydrogen {5-ethyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol chloride
Synonyms: dihydroquinidine
Molecular Weight (g/mol): 362.9
Molecular Formula: C20H27ClN2O2
InChi Key: MULXTQKDWYBJMO-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCC1CN2CCC1CC2C(O)C1=C2C=C(OC)C=CC2=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results