accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1477-55-0

IUPAC Name: 1-[3-(aminomethyl)phenyl]methanamine
Synonyms: M-xylylenediamine 1,3-benzenedimethanamine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: FDLQZKYLHJJBHD-UHFFFAOYSA-N
SMILES: NCC1=CC(CN)=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)