Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylylenediamine 99.0+%, TCI America™
SDP

Catalog No. D0127500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0127500G 500 g
D012725G 25 g
D0127100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0127500G Supplier TCI America Supplier No. D0127500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1477-55-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00008119
InChI Key FDLQZKYLHJJBHD-UHFFFAOYSA-N
Synonym m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis
PubChem CID 15133
IUPAC Name [3-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)CN

Specifications

Color Yellow
Boiling Point 248°C
Quantity 500 g
UN Number 2735
Formula Weight 136.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Xylylenediamine
EINECSNumber (3)-0308
RTECSNumber PF8970000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.