accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNONSOONHOSCH3FFF

CAS RN 147776-06-5

IUPAC Name: N-{[4'-({3-butyl-5-oxo-1-[2-(trifluoromethyl)phenyl]-4,5-dihydro-1H-1,2,4-triazol-4-yl}methyl)-[1,1'-biphenyl]-2-yl]sulfonyl}-3-methylthiophene-2-carboxamide
Synonyms: N-[4'-({3-butyl-5-oxo-1-[2-(trifluoromethyl)phenyl]-1,2,4-triazol-4-yl}methyl)-[1,1'-biphenyl]-2-ylsulfonyl]-3-methylthiophene-2-carboxamide
Molecular Weight (g/mol): 654.72
Molecular Formula: C32H29F3N4O4S2
InChi Key: MMDNKTXNUZFVKD-UHFFFAOYSA-N
SMILES: CCCCC1=NN(C(=O)N1CC1=CC=C(C=C1)C1=CC=CC=C1S(=O)(=O)NC(=O)C1=C(C)C=CS1)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)