accessibility menu, dialog, popup

UserName

Viewing profile as user:
IH3CNOONH3C(Z)(E)(E/Z)

CAS RN 14806-50-9

IUPAC Name: 3-ethyl-2-[(3E)-5-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzoxazol-3-ium iodide
Synonyms: 3-ethyl-2-[(3E)-5-[(2Z)-3-ethyl-1,3-benzoxazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzoxazol-3-ium iodide
Molecular Weight (g/mol): 486.35
Molecular Formula: C23H23IN2O2
InChi Key: CLDZYSUDOQXJOU-UHFFFAOYSA-M
SMILES: [I-].CCN1\C(OC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CC)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Color 
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)