Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diethyloxadicarbocyanine Iodide, Laser Grade 99%, Thermo Scientific™

Catalog No. AC407365000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407365000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407365000 Supplier Thermo Scientific Chemicals Supplier No. 407365000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14806-50-9
Molecular Formula C23H23IN2O2
Molecular Weight (g/mol) 486.35
MDL Number MFCD00011953,MFCD00011953
InChI Key CLDZYSUDOQXJOU-UHFFFAOYSA-M
Synonym DODC iodide
PubChem CID 91972131
ChEBI CHEBI:51899
IUPAC Name 3-ethyl-2-[(3E)-5-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzoxazol-3-ium iodide
SMILES [I-].CCN1\C(OC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CC)C2=CC=CC=C2O1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Fluorescence Max 605nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 578nm (in methanol)
Formula Weight 486.34
Molar Absorption Coefficient 256000L/(mole.cm) (lambda max.)
Suitability (for use as a laser dye) passes
UV_Vis 98.5% min.
Color Blue to Purple
Quantity 500 mg
Physical Form Crystalline Powder
Chemical Name or Material 3, 3'-Diethyloxadicarbocyanine iodide, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.