Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diethyloxadicarbocyanine iodide, 96%

Catalog No. AAH33094MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33094MD 250 mg
AAH3309403 1 g
AAH3309406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33094MD Supplier Thermo Scientific Chemicals Supplier No. H33094MD
  • $58.30
    $46.65 / Each of 1

    Save $11.65

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14806-50-9 | C23H23IN2O2 | 486.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14806-50-9
Molecular Formula C23H23IN2O2
Molecular Weight (g/mol) 486.35
MDL Number MFCD00011953,MFCD00011953
InChI Key CLDZYSUDOQXJOU-UHFFFAOYSA-M
Synonym dodc iodide, 3-ethyl-2-5-3-ethyl-2-benzoxazolinylidene-1,3-pentadienyl benzoxazolium iodide, 5-3-ethylbenzoxazol-2-ylidene-1-3-ethylbenzoxazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972131
ChEBI CHEBI:51899
IUPAC Name (2Z)-3-ethyl-2-[5-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzoxazole;iodide
SMILES [I-].CCN1\C(OC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CC)C2=CC=CC=C2O1

Specifications

Melting Point ∽215°C (decomposition)
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 486.36
Percent Purity 96%
Chemical Name or Material 3,3'-Diethyloxadicarbocyanine iodide
Hazard Category H300
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 238-873-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.