accessibility menu, dialog, popup

UserName

Viewing profile as user:
F

CAS RN 1481-36-3

IUPAC Name: fluorocyclopentane
Synonyms: fluorocyclopentane
Molecular Weight (g/mol): 88.13
Molecular Formula: C5H9F
InChi Key: YHYNFMGKZFOMAG-UHFFFAOYSA-N
SMILES: FC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results