Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorocyclopentane 95.0+%, TCI America™
SDP

Catalog No. F05885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05885G Supplier TCI America Supplier No. F05885G

Chemical Identifiers

CAS 1481-36-3
Molecular Formula C5H9F
Molecular Weight (g/mol) 88.125
MDL Number MFCD08059532
InChI Key YHYNFMGKZFOMAG-UHFFFAOYSA-N
PubChem CID 137022
IUPAC Name fluorocyclopentane
SMILES C1CCC(C1)F

Specifications

Color Yellow
Boiling Point 52°C
Quantity 5 g
UN Number 1993
Formula Weight 88.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Fluorocyclopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.