accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiSHOH3COH3C

CAS RN 14814-09-6

IUPAC Name: 3-(triethoxysilyl)propane-1-thiol
Synonyms: 3-(triethoxysilyl)propane-1-thiol
Molecular Weight (g/mol): 238.42
Molecular Formula: C9H22O3SSi
InChi Key: DCQBZYNUSLHVJC-UHFFFAOYSA-N
SMILES: CCO[Si](CCCS)(OCC)OCC
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)