Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Mercaptopropyl)triethoxysilane 96.0+%, TCI America™
SDP

Catalog No. M1505100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1505100G 100 g
M150525G 25 g
M1505500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1505100G Supplier TCI America Supplier No. M1505100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14814-09-6
Molecular Formula C9H22O3SSi
Molecular Weight (g/mol) 238.417
MDL Number MFCD00053924
InChI Key DCQBZYNUSLHVJC-UHFFFAOYSA-N
Synonym 3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy
PubChem CID 84658
IUPAC Name 3-triethoxysilylpropane-1-thiol
SMILES CCO[Si](CCCS)(OCC)OCC

Specifications

Color Yellow
Boiling Point 115°C
Quantity 100 g
Formula Weight 238.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Mercaptopropyl)triethoxysilane
EINECSNumber (2)-2045
RTECSNumber TZ7760000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.