accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO

CAS RN 1483-67-6

IUPAC Name: hept-2-ynoic acid
Synonyms: hept-2-ynoic acid
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: UEERQXQKEJPYBR-UHFFFAOYSA-N
SMILES: CCCCC#CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results