accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1483-73-4

IUPAC Name: diphenyliodanium bromide
Synonyms: diphenyliodanium bromide
Molecular Weight (g/mol): 361.02
Molecular Formula: C12H10BrI
InChi Key: LGPSGXJFQQZYMS-UHFFFAOYSA-M
SMILES: [Br-].[I+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)