Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium bromide, 98%, Thermo Scientific™

Catalog No. AAA1299609
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1299609 Supplier Thermo Scientific Chemicals Supplier No. A1299609

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1483-73-4
Molecular Formula C12H10BrI
Molecular Weight (g/mol) 361.02
MDL Number MFCD00031703
InChI Key LGPSGXJFQQZYMS-UHFFFAOYSA-M
Synonym diphenyliodonium bromide, iodonium, diphenyl-, bromide, diphenyliodanium bromide, iodonium, diphenyl-, bromide 1:1, diphenyliodoniumbromide, diphenyl iodonium bromide, acmc-1c252, iodonium, diphenyl-,bromide 1:1
PubChem CID 73871
IUPAC Name diphenyliodanium bromide
SMILES [Br-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼244°C (sublimation)
Odor Odorless
Quantity 10 g
UN Number UN2811
Beilstein 3918437
Formula Weight 361.03
Percent Purity 98%
Chemical Name or Material Diphenyliodonium bromide
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.