Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium Bromide 98.0+%, TCI America™
SDP

Catalog No. D23725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23725G Supplier TCI America Supplier No. D23725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1483-73-4
Molecular Formula C12H10BrI
Molecular Weight (g/mol) 361.02
MDL Number MFCD00031703
InChI Key LGPSGXJFQQZYMS-UHFFFAOYSA-M
Synonym diphenyliodonium bromide, iodonium, diphenyl-, bromide, diphenyliodanium bromide, iodonium, diphenyl-, bromide 1:1, diphenyliodoniumbromide, diphenyl iodonium bromide, acmc-1c252, iodonium, diphenyl-,bromide 1:1
PubChem CID 73871
IUPAC Name diphenyliodanium bromide
SMILES [Br-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 188°C
Quantity 5 g
UN Number 2811
Formula Weight 361.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.