accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14836-60-3

IUPAC Name: mercury(1+) nitric acid dihydrate
Synonyms: mercury(1+) nitric acid dihydrate
Molecular Weight (g/mol): 299.63
Molecular Formula: H5HgNO5
InChi Key: GJKBHHZLGNEESF-UHFFFAOYSA-N
SMILES: O.O.[Hg+].O[N+]([O-])=O
Grade 
  • (1)
  • (5)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (9)
  • (1)
  • (5)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)