Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercurous Nitrate, Purified, Spectrum™ Chemical
SDP

Catalog No. 18611642 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
110 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-642 12 kg Poly Pail
18-611-640 110 lb. Fiber Drum
18-611-641 125 g Amber Glass Bottle
18-611-643 500 g Amber Glass Bottle
18-611-639 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-642 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M120612KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14836-60-3

Specifications

CAS Max % 1
Quantity 12 kg
Molecular Weight (g/mol) 525.192
Grade Purified
Packaging Poly Pail
Chemical Name or Material Mercurous Nitrate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Caution: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.