Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercurous Nitrate ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK1434500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-1434-500 500 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-1434-500 Supplier Avantor Supplier No. 143404

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14836-60-3
Molecular Formula Hg2(NO3)2·2H2O

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 561.22
Grade ACS
Packaging Glass
Chemical Name or Material Mercurous Nitrate, Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.