CAS RN 148642-42-6
IUPAC Name:
N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]-2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-carboxamide hydrochloride
Synonyms:
GR 127935 hydrochloride GR127935 hydrochloride
Molecular Weight (g/mol):
534.06
Molecular Formula:
C29H32ClN5O3
InChi Key:
SRVVUYIJVBLEJI-UHFFFAOYSA-N
SMILES:
Cl.COC1=C(C=C(NC(=O)C2=CC=C(C=C2)C2=C(C)C=C(C=C2)C2=NOC(C)=N2)C=C1)N1CCN(C)CC1
1
–
1
of
1
results
1
R&D Systems™ GR 127935 hydrochloride
CAS: 148642-42-6 Molecular Formula: C29H32ClN5O3