accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClHO

CAS RN 14868-03-2

IUPAC Name: 4-[2,2-dichloro-1-(4-hydroxyphenyl)ethenyl]phenol
Synonyms: dihydroxymethoxychlor olefin
Molecular Weight (g/mol): 281.13
Molecular Formula: C14H10Cl2O2
InChi Key: OWEYKIWAZBBXJK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=C(Cl)Cl)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results