accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CONNCH3(S)OCH3OH3COOH3CH

CAS RN 148849-67-6

IUPAC Name: hydrogen 3-[3-({[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl}(methyl)amino)propyl]-7,8-dimethoxy-2,3,4,5-tetrahydro-1H-3-benzazepin-2-one chloride
Synonyms: hydrogen ivabradine chloride
Molecular Weight (g/mol): 505.05
Molecular Formula: C27H37ClN2O5
InChi Key: HLUKNZUABFFNQS-ZMBIFBSDSA-N
SMILES: [H+].[Cl-].COC1=CC2=C(CC(=O)N(CCCN(C)C[C@H]3CC4=CC(OC)=C(OC)C=C34)CC2)C=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)