accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CO(S)O(S)H3COCH3OHOHHNO(S)CH3(S)OH(R)CH3(R)OH(R)CH3(S)OCH3O(R)CH3CH3HOO(?)(Z)(Z)

CAS RN 14897-39-3

IUPAC Name: sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
Synonyms: sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
Molecular Weight (g/mol): 720.77
Molecular Formula: C37H47NNaO12
InChi Key: YVOFSHPIJOYKSH-DITFEXNXSA-N
SMILES: [Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O
Quantity 
  • (2)
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results