Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifamycin SV Sodium Salt, MP Biomedicals™

Catalog No. MP021915162 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021915162 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021915162 Supplier MP Biomedicals, Inc Supplier No. 02191516.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to study potential genes that may be involved in the conversion of rifamycin SV into rifamycin B. Rifamycin SV Sodium Salt, MP Biomedicals is a broad-spectrum semisynthetic antimicrobial agent of the rifamycin group. It has limited oral absorption.

Rifamycin SV inhibits selective (E. coli, B. subtilis) bacterial DNA-dependent RNA polymerase by binding to the polymerase β-subunit, a mechanism similar to rifabutin. It acts as a selective cytochrome P450 3A4 inducer. It has limited oral absorption. It is active against Gram-positive bacteria and is moderately active against Gram-negative organisms.

  • Used to study potential genes, such as rif15 and rif16, which may be involved in the conversion of rifamycin SV into rifamycin B
  • Potential treatment for traveler’s diarrhea (TD), Clostridium difficile-associated disease (CDAD) and rheumatoid knee synovitis
  • It is used in electron spin resonance studies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14897-39-3
Molecular Formula C37H47NNaO12
Molecular Weight (g/mol) 720.77
MDL Number MFCD09752825
InChI Key YVOFSHPIJOYKSH-DITFEXNXSA-N
Synonym rifamycin sodium salt, rifamycin sodium usan, rifamycin sv sodium salt, rifocina tn, otofa tn
PubChem CID 124219641
IUPAC Name sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
SMILES [Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O

Specifications

Quantity 250 mg
Solubility Information Ethanol: soluble (50mg/mL)
Formula Weight 719.8
Grade Not Available
Physical Form Powder
Recommended Storage +4°C. Store under nitrogen. Protect from light.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.