Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifamycin SV sodium salt, MP Biomedicals™

Catalog No. ICN19151605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19151605 5 g
ICN19151601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19151605 Supplier MP Biomedicals, Inc Supplier No. 0219151605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14897-39-3
Molecular Formula C37H47NNaO12
Molecular Weight (g/mol) 720.77
MDL Number MFCD09752825
InChI Key YVOFSHPIJOYKSH-DITFEXNXSA-N
Synonym rifamycin sodium salt, rifamycin sodium usan, rifamycin sv sodium salt, rifocina tn, otofa tn
PubChem CID 124219641
IUPAC Name sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
SMILES [Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O

Specifications

Color Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material Rifamycin SV sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.