CAS RN 149022-15-1
3-Methyl-2-Benzothiazolinone Hydrazone Hydrochloride, Hydrate, Spectrum™ Chemical
CAS: 149022-15-1
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
CAS: 149022-15-1 Molecular Formula: C8H10ClN3S Molecular Weight (g/mol): 215.70 MDL Number: MFCD00149370 InChI Key: OEZPVSPULCMUQB-UHFFFAOYSA-N Synonym: unii-9u2c4w7epr,mbth hydrochloride,9u2c4w7epr,3-methyl-2-benzothiazolone hydrazone hydrochloride,3-methyl-2-benzothiazolinone hydrazone hydrochloride,z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride,mbth hcl,ccris 1543,2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1,2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride PubChem CID: 9575839 IUPAC Name: (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride SMILES: Cl.CN1C(SC2=CC=CC=C12)=NN
3-Methyl-2-benzothiazolinonehydrazone Hydrochloride Hydrate 98.0+%, TCI America™
CAS: 149022-15-1 Molecular Formula: C8H10ClN3S Molecular Weight (g/mol): 215.70 MDL Number: MFCD00149370 InChI Key: OEZPVSPULCMUQB-UHFFFAOYSA-N Synonym: unii-9u2c4w7epr,mbth hydrochloride,9u2c4w7epr,3-methyl-2-benzothiazolone hydrazone hydrochloride,3-methyl-2-benzothiazolinone hydrazone hydrochloride,z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride,mbth hcl,ccris 1543,2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1,2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride PubChem CID: 9575839 IUPAC Name: 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride SMILES: Cl.CN1C(SC2=CC=CC=C12)=NN
MBTH Indicator, 0.05% (w/v), Ricca Chemical
Molecular Formula: C8H10ClN3S Molecular Weight (g/mol): Mixture InChI Key: OEZPVSPULCMUQB-UHFFFAOYSA-N PubChem CID: 9575839 IUPAC Name: 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride SMILES: Cl.CN1C(SC2=CC=CC=C12)=NN
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride TS, 0.1% (w/v) in 90% (v/v) Methanol, Ricca Chemical
CAS: 149022-15-1 Molecular Formula: C8H10ClN3S Molecular Weight (g/mol): 215.70 MDL Number: MFCD00004595 InChI Key: OEZPVSPULCMUQB-UHFFFAOYSA-N Synonym: unii-9u2c4w7epr,mbth hydrochloride,9u2c4w7epr,3-methyl-2-benzothiazolone hydrazone hydrochloride,3-methyl-2-benzothiazolinone hydrazone hydrochloride,z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride,mbth hcl,ccris 1543,2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1,2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride IUPAC Name: 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride SMILES: Cl.CN1C(SC2=CC=CC=C12)=NN