accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBrOHO

CAS RN 14903-90-3

IUPAC Name: 3-bromofuran-2-carboxylic acid
Synonyms: 3-bromofuran-2-carboxylic acid
Molecular Weight (g/mol): 190.98
Molecular Formula: C5H3BrO3
InChi Key: UZBGSJZFBUOJNE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results