accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 149082-03-1

CH3NOHH3CNOO

CAS RN 149082-03-1

IUPAC Name: (3,5-dimethyl-4-nitropyridin-2-yl)methanol
Synonyms: (3,5-dimethyl-4-nitropyridin-2-yl)methanol
Molecular Weight (g/mol): 182.18
Molecular Formula: C8H10N2O3
InChi Key: KBCDOXSSYLFMHH-UHFFFAOYSA-N
SMILES: CC1=CN=C(CO)C(C)=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results