accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14913-33-8

IUPAC Name: dichloroplatinum diamine
Synonyms: cisplatin
Molecular Weight (g/mol): 300.05
Molecular Formula: Cl2H6N2Pt
InChi Key: LXZZYRPGZAFOLE-UHFFFAOYSA-L
SMILES: N.N.Cl[Pt]Cl
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results